7X™ Concentrate, MP Biomedicals
Supplier: MP Biomedicals
097667093 097667094
IC7667093EA 96.34 USD
IC7667093 IC7667094
7X™ Concentrate, MP Biomedicals
Cleaning Agents Detergents and Surface Cleansers
This concentrated solution was designed to safely clean laboratory plastics and glassware simply by soaking overnight
You may boil equipment in 7X™. This product’s surfactants are extremely effective and powerful.
Order Now
SPECIFICATIONS
Environmentally Preferable
pH 9.0 - 11.0