N-Lauroylsarcosine sodium salt ≥94%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Lauroylsarcosine sodium, Sarkosyl, Sodium lauroyl sarcosine, Sodium N-lauroylsarcosinate, Sodium N-dodecanoyl-N-methylglycinate

0219028950 0219028980 0219028990
IC19028950EA 123.94 USD
IC19028950 IC19028980 IC19028990
N-Lauroylsarcosine sodium salt ≥94%, white powder
N-Lauroylsarcosine sodium salt

N-Lauroylsarcosine is an anionic detergent.


N-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoproteins; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungistatic agent in aqueous dispersion (1%).


Sodium Salt: Soluble in water (100 mg/mL - clear to slightly hazy, colorless solution).


N-Lauroylsarcosine is an anionic detergent.

Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
Storage Temperature: Ambient
MDL Number: MFCD00042728
CAS Number: 137-16-6
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e15248912e48b73e2c646c8186e92ab3c8da7ba&vid=73c9ba74fd6fee332c7b7037d139aceb3b92dd00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥94%
pH (10% aqueous solution) 7,5 - 8,5
Aggregation Number 2
CMC 14,57 mM at 30 °C (Lit.)
Extinction Coefficient EM = 3 (280 nm) (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR