Carbol fuchsin, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride

0215493310
IC15493310EA 109.66 USD
IC15493310
Carbol fuchsin, powder
Carbol fuchsin

Soluble in ethanol, water.


Carbol Fuchsin is a mixture of basic fuchsin and phenol.

Formula: C₂₁H₂₂ClN₃
MDL Number: MFCD00013129
CAS Number: 4197-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR