Benzalkonium chloride ≥80%, viscous solution

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride

0215043083 0215043091
IC15043083EA 79.68 USD
IC15043083 IC15043091
Benzalkonium chloride ≥80%, viscous solution
Benzalkonium chloride

It has been reported that benzalkonium chloride inactivates HIV and HSV in vitro.


It is not effective against gram-negative bacteria or picornaviruses and it is not recommended for treatment of sanitising critical or semicritical medical instruments. Incompatible with anionic detergents, such as soap, and with nitrates. The preservative effect can be neutralized by the addition of Tween 80 and lecithin.


Soluble in water, alcohol, acetone; slightly soluble in benzene. Aqueous solutions are slightly alkaline to litmus and foams strongly when shaken. Composed of C12, C14, and C16 alkylbenzyldimethylammonium chlorides. Approximate ratios: C12 (40%) : C14 (50%) : C16 (10%).


Caution: Research Use Only (RUO).

Formula: C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
Storage Temperature: Ambient
MDL Number: MFCD00145757
CAS Number: 63449-41-2
UN: 2923
ADR: 8 III

Order Now

Specification Test Results

Identity Test Passes
Concentration 80% Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR