FastPrep™ Metal Lysing Matrix Tubes, MP Biomedicals
Supplier: MP Biomedicals
116991002 116991003 116991006 116992002 116992003 116992006
IC116991002PK 107.85 USD
IC116991002 IC116991003 IC116991006 IC116992002 IC116992003 IC116992006
FastPrep™ Metal Lysing Matrix Tubes, MP Biomedicals
Homogenizers Tissue Grinders
Stainless Steel Tubes are ideal for grinding, lysis, and homogenization of the most resistant samples.
  • Dry grinding very tough or hard samples where heat generated can damage plastic tubes
  • Cryogenic dry grinding where severe cold temps (dry ice or LN2) can damage plastic tubes
  • Milling or grinding non-biological samples where plastic contamination is a concern
  • Sample processing with solvents or chemicals that are incompatible with plastics
Constructed from 308 stainless steel, these tubes and grinding matrix are tough enough to stand up to the mostMore Product Information