1X Hanks' Balanced Salt Solution (Modified) without Phenol Red, MP Biomedical
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
091810254
IC091810254EA 41.49 USD
IC091810254
1X Hanks' Balanced Salt Solution (Modified) without Phenol Red, MP Biomedical
Buffers Cell Culture Buffers
Phenol red has been shown to interfere with the growth of some cells at cloning densities. Use this medium when working with stem cells or when growing cells at low densities.
  • Stem cells growth
  • Growth of cells at low densities