Adenosine 99.0-101.0% USP

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside

0219980305 0219980325 0219980380 0219980391
IC0219980305EA 49.29 USD
IC0219980305 IC0219980325 IC0219980380 IC0219980391
Adenosine 99.0-101.0% USP
Adenosine

Adenosine is a purine nucleoside comprising a molecule of adenine attached to a ribose sugar molecule moiety via a β-N9-glycosidic bond. It plays an important role in energy transfer. It is an endogenous purine nucleoside that modulates physiological process wherein cellular signaling by adenosine occurs through four known adenosine receptor subtypes (A1, A2A, A2B, and A3) Adenosine is an anti-inflammatory agent at the A(2A) receptor. Cardioprotective effects may relate to activation of A1 adenosine receptors. Endogenous neurotransmitter at adenosine receptors. Cardioprotective effects may relate to activation of A1 adenosine receptors. The antiplatelet and anti−inflammatory actions of adenosine appear to be mediated via the A2 adenosine receptor. In contrast, adenosine appears to be a pro-inflammatory mediator in asthma and chronic obstructive pulmonary disease (COPD). Adenosine is used in topical treatment of foot wounds in diabetes mellitus and also used for the treatment or prevention of cardiac arrhythmias (irregular heartbeat).It is used to cause dilation of the blood vessels. Physical Appearance: White powder Optical Rotation: -68.0° to -72.0° (c=2, NaOH 1:20) Heavy Metals: ≤0.001% Loss on Drying: ≤0.5% Residue on Ignition: ≤0.1% Storage Temp. : -20 °C

Formula: C₁₀H₁₃N₅O₄
Melting Pt: 233…238 °C
Storage Temperature: Freezer
MDL Number: MFCD00005752
CAS Number: 58-61-7
EINECS: 200-389-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=690072dfe7e9273ef6c19d75d34d84ba711a0082&vid=690072dfe7e9273ef6c19d75d34d84ba711a0082&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR