STS 45i DNA Sequencing Unit, IBI Scientific
Supplier: IBI Scientific
IB80000
IB80000EA 1856.03 USD
IB80000
STS 45i DNA Sequencing Unit, IBI Scientific
Electrophoresis Sequencers
Slant-back design holds glass plates in place during assembly
Built-in aluminum thermoplate distributes heat evenly, preventing gel artifacts. White backing of the thermoplate allows easy viewing of dye migrations. Epoxy coating makes thermoplate electrically inert for increased user safety. Modular platinum electrodes are recessed. The lower buffer tank is removable for easy cleaning and disposal of radioactive material. Upper reservoir has a drainage port for easy drainage of buffer into a sink or separate container. Both chambers feature a hinged, interlocking safety lid that allowsMore Product Information