p-Anisaldehyde zS, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 4-Methoxybenzaldehyde

MQ200

8.22314.0250
EM8.22314.0250CS 644 USD
EM8.22314.0250
p-Anisaldehyde zS, Sigma-Aldrich®
p-Anisaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 140
Melting Pt: 0…2 °C
Density: 1.12 g/cm³ (25 °C)
Flash Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00003385
CAS Number: 123-11-5
EINECS: 204-602-6
Merck Index: 13,00666

Order Now

Specification Test Results

Solubility 2 g/l (20 °C)
Molar Mass 136.15 g/mol
Boiling Point 247 - 249 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 116 °C
Refractive Index 1.57 (20 °C)
Explosion Limit 1.4 - 5.3 %(V)
Density 1.12 g/cm3 (25 °C)
pH 7 (2 g/l, H2O, 20 °C)
Ignition Point 220 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR