Tween® 20 (Polysorbate) zS, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Polyoxyethylene sorbitan monolaurate

MQ200

8.22184.2500 8.22184.0500
EM8.22184.2500EA 375.93 USD
EM8.22184.2500 EM8.22184.0500
Tween® 20 (Polysorbate) zS, Sigma-Aldrich®
Tween® 20 (Polysorbate)
Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now

Specification Test Results

Solubility (25 °C) soluble
Molar Mass 1228 g/mol
Boiling Point 100 °C
Vapor Pressure  - 1.4 hPa (20 °C)
Flash Point 149 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR