Lithium iodide, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.18287.0050
EM8.18287.0050CS 16313 USD
EM8.18287.0050
Lithium iodide, anhydrous for synthesis, Sigma-Aldrich®
Lithium iodide
Formula: ILi
MDL Number: MFCD00011092
CAS Number: 10377-51-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 450 °C
Molar Mass 133.84 g/mol
Bulk Density 1000 kg/m3
Boiling Point 1180 °C
Density 4 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR