L-Methionine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid

M-Clarity™ quality level = MQ200

8.16010.0025 8.16010.0100
EM8.16010.0025CS 5156 USD
EM8.16010.0025 EM8.16010.0100
L-Methionine for synthesis, Sigma-Aldrich®
L-Methionine
Formula: C₅H₁₁NO₂S
MDL Number: MFCD00063097
CAS Number: 63-68-3

Order Now

Specification Test Results

Solubility 48 g/l (20 °C)
Melting Point 280 - 285 °C
Molar Mass 149.2 g/mol
Bulk Density 550 kg/m3
pH 5 - 7 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR