Triphenylphosphine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.08270.0005 8.08270.0250 8.08270.1000
EM8.08270.0005CS 36462 USD
EM8.08270.0005 EM8.08270.0250 EM8.08270.1000
Triphenylphosphine for synthesis, Sigma-Aldrich®
Triphenylphosphine
Formula: C₁₈H₁₅P
Boiling Pt: 377 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.194 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0
Merck Index: 12,09873

Order Now

Specification Test Results

Solubility  - 0.0001 g/l (25 °C)
Melting Point 78.5 - 81.5 °C
Molar Mass 262.29 g/mol
Bulk Density 500 - 600 kg/m3
Boiling Point 195 - 205 °C (7 hPa)
Vapor Pressure 0.01 hPa (88 °C)
Flash Point 182 °C
Density 1.194 g/cm3 (20 °C)
Ignition Point 425 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR