Polyethylene glycol 400, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: PEG 400

M-Clarity™ quality level = MQ200

8.07485.1000
EM8.07485.1000EA 72 USD
EM8.07485.1000
Polyethylene glycol 400, Sigma-Aldrich®
Polyethylene glycol 400
Formula: H(OCH₂CH₂)nOH
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 4…8 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 245 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR