LiChroTest® Standard samples for HPLC system qualification Set 2: Precision; 100 mg/L Methyl paraben in methanol / water (50:50), MilliporeSigma
Supplier: MilliporeSigma
1.19158.0001
EM1.19158.0001CS 38978 USD
EM1.19158.0001
LiChroTest® Standard samples for HPLC system qualification Set 2: Precision; 100 mg/L Methyl paraben in methanol / water (50:50), MilliporeSigma
Standards Chromatography Standards