MColortest™ Nitrite Test In Freshwater And Seawater, MilliporeSigma
Supplier: MilliporeSigma
1.14658.0001
EM1.14658.0001EA 83 USD
EM1.14658.0001
MColortest™ Nitrite Test In Freshwater And Seawater, MilliporeSigma
Environmental Test Kits
Nitrite is naturally present in soil, water and food, and regularly testing its concentration is essential for many applications
The test kit measures nitrite ions in freshwater and contains a colour card for the visual comparison.