Acetonitrile for DNA synthesis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

M-Clarity™ quality level = MQ200

1.13212.2500 1.13212.4000
EM1.13212.2500CS 1127 USD
EM1.13212.2500 EM1.13212.4000
Acetonitrile for DNA synthesis, Supelco®
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -45.7 °C
Molar Mass 41.05 g/mol
Boiling Point 81.6 °C (1013 hPa)
Vapor Pressure 97 hPa (20 °C)
Flash Point 2 °C
Refractive Index 1.34
Explosion Limit 3.0 - 17 %(V)
Density 0.786 g/cm3 (20 °C)
pH (H2O) no data available
Ignition Point 524 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR