TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) analytical reagent, Millipore®

Supplier: MilliporeSigma
Danger

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

M-Clarity™ quality level = MQ200

1.08603.1000 1.08603.2500
EM1.08603.1000EA 150 USD
EM1.08603.1000 EM1.08603.2500
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) analytical reagent, Millipore®
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)

Triton® X-100 for analysis.

Formula: C₈H₁₇C₆H₄(OCH₂CH₂)⁷nOH
Boiling Pt: >200 °C (1013 hPa)
Melting Pt: 6
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 251 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9 III
Merck Index: 13,06793

Order Now

Specification Test Results

Identity (IR spectrum) passes test
pH value (10 g/l, H₂O, 20 °C) 5.0 - 8.0
Density (d 20/4) 1.064 - 1.067
Cloud point 63 - 69 °C
Ethylene oxide (GC) ≤30 ppm
Dioxane (GC) ≤20 ppm
Sulphated ash (600 °C) ≤0.2%
Water (acc. to Karl Fischer) ≤0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR