Pararosaniline hydrochloride, Certistain® Microscopy (C.I. 42500), Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Basic Parafuchsin, 4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride, [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride, Basic Fuchsin hydrochloride, Parafuchsin hydrochloride, Pararosaniline chloride, Basic Red 9, 4,4'-((4-Iminocyclohexa-2,5-dien-1-ylidene)methylene)dianiline hydrochloride, Paramagenta hydrochloride

1.07509.0100
EM1.07509.0025CS 11485.1 USD
EM1.07509.0025 EM1.07509.0100
Pararosaniline hydrochloride, Certistain® Microscopy (C.I. 42500), Sigma-Aldrich®
Pararosaniline hydrochloride

The PAS (periodic acid Schiff) reaction is one of the most frequently used chemical methods for histology. This Certistain® Pararosaniline (chloride) solid dye is used to prepare the Schiff’s reagent required for the PAS reaction, for the staining of mucopolysaccharides in histological tissue specimens.
As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations.

Formula: C₁₉H₁₈ClN₃
MW: 323.82 g/mol
Melting Pt: 268
MDL Number: MFCD00001657
CAS Number: 569-61-9

Order Now

Specification Test Results

Solubility 10 g/l (25 °C)
Melting Point 268 - 270 °C decomposes
Molar Mass 323.83 g/mol
Bulk Density 500 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR