Trisodium phosphate dodecahydrate ≥93.5% (by alkalimetry), EMPROVE® ESSENTIAL E339, NF, SAFC®

Supplier: MilliporeSigma
Warning

Synonyms: Sodium Phosphate tribasic 12-Hydrate, Sodium phosphate dodecahydrate, Sodium phosphate tribasic dodecahydrate, Trisodium phosphate

1.06577.5000
EM1.06577.5000CS 1530.78 USD
EM1.06577.5000
Trisodium phosphate dodecahydrate ≥93.5% (by alkalimetry), EMPROVE® ESSENTIAL E339, NF, SAFC®
tri-Sodium phosphate dodecahydrate
Formula: Na₃PO₄·12H₂O
Melting Pt: ∼75 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149198
CAS Number: 10101-89-0
EINECS: 231-509-8
REACH: 01-2119489800-32-xxxx
Merck Index: 13,08736

Order Now

Specification Test Results

Identity passes test
Assay (alkalimetric, calculated on calcined substance) ≥ 93.5 %
Free alkali (as NaOH) ≤ 2.5 %
In water insoluble matter (calculated on dried substance) ≤ 0.2 %
pH-value (1 %, water) 11.5 - 12.5
Fluoride (F) ≤ 0.0010 %
Heavy metals (as Pb) ≤ 0.0010 %
As (Arsenic) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.0001 %
Hg (Mercury) ≤ 0.0001 %
Pb (Lead) ≤ 0.0001 %
Loss on ignition (800 °C) 45.0 - 57.0 %
Residual solvents (ICH Q3C) excluded by production process
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to NF, E 339
Conforms to the purity criteria on food additives according to the current European Commission Regulation.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR