Methanol-[D₄] ≥99.8% (Isotopic) 99.8, MagniSolv™ for NMR spectroscopy for NMR spectroscopy, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol

1.06028.0005 1.06028.0009 1.06028.0010 1.06028.0025 1.06028.0100
EM1.06028.0001EA 38 USD
EM1.06028.0001 EM1.06028.0005 EM1.06028.0009 EM1.06028.0010 EM1.06028.0025 EM1.06028.0100
Methanol-[D₄] ≥99.8% (Isotopic) 99.8, MagniSolv™ for NMR spectroscopy for NMR spectroscopy, Sigma-Aldrich®
Methanol-[D4]

MagniSolv™ deuterated solvents provide:
• Reliable results in the NMR-spectra by excellent chemical purity and highest isotopic enrichment.
• Reliable deuteration degrees.
• Low residual water content.
• Determination of water content in two ways.

Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 65.3
Melting Pt: -99
Density: 0.888 g/cm³ (20 °C)
Flash Pt: 11
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3 (6.1) II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR