Potassium nitrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Nitrate of Potash

1.05063.0500 1.05063.1000 1.05063.5000 1.05063.9025
EM1.05063.0500CS 593 USD
EM1.05063.0500 EM1.05063.1000 EM1.05063.5000 EM1.05063.9025
Potassium nitrate, EMSURE® for analysis, Supelco®
Potassium nitrate
Formula: KNO₃
Melting Pt: 334 °C
Density: 2.109 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733

Order Now

Specification Test Results

Solubility 320 g/l (20 °C)
Melting Point 334 °C
Molar Mass 101.10 g/mol
Bulk Density 800 kg/m3
Density 2.11 g/cm3 (20 °C)
pH 5.0 - 7.5 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR