n-Hexane ≥98.50% (hexane isomers + methylcyclopentane, GC), ≥96.0% (GC)] [≥96.0% (GC)], EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

M-Clarity™ quality level = MQ300

1.04374.1000 1.04374.2500 1.04374.2511 1.04374.4000 1.04374.6025
EM1.04374.1000CS 582 USD
EM1.04374.1000 EM1.04374.2500 EM1.04374.2511 EM1.04374.4000 EM1.04374.6025
n-Hexane ≥98.50% (hexane isomers + methylcyclopentane, GC), ≥96.0% (GC)] [≥96.0% (GC)], EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
n-Hexane
Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C (lit.)
Melting Pt: –95 °C (lit.)
Density: 0.659 g/ml (25°C) (lit.)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥ 96.0 %
Purity Σ hexane isomers + methylcyclopentane (GC) ≥ 98.5 %
Identity (IR) conforms
Color ≤ 10 Hazen
Water-soluble titrable acid ≤ 0.0003 meq/g
Refractive index (n 20/D) 1.375 - 1.376
Density (d 20 °C/20 °C) 0.659 - 0.663
Boiling range (min.95 %) 67 - 69 °C
Transmission (between 260 nm and 420 nm) ≥ 97 %
Thiophene conforms
Aromatics (as benzene) ≤ 0.01 %
Sulfur compounds (as S) ≤ 0.005 %
Readily carbonizable substances conforms
Al (Aluminium) ≤ 0.00005 %
B (Boron) ≤ 0.000002 %
Ba (Barium) ≤ 0.00001 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Sn (Tin) ≤ 0.00001 %
Zn (Zinc) ≤ 0.00001 %
Evaporation residue ≤ 0.001 %
Water ≤ 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR