n-Hexane ≥95% (by GC), EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

M-Clarity™ quality level = MQ200

1.04368.1000 1.04368.2500 1.04368.2511 1.04368.6025 1.04368.9011 1.04368.9190
EM1.04368.1000CS 402 USD
EM1.04368.1000 EM1.04368.2500 EM1.04368.2511 EM1.04368.6025 EM1.04368.9011 EM1.04368.9190
n-Hexane ≥95% (by GC), EMPLURA®, Supelco®
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 (lit.)
Melting Pt: -94.3 (lit.)
Density: 0.659 g/ml (25°C) (lit.)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR