Iron(II) sulfate hydrate 86-89% (by manganometry) FCC, BP, USP, SAFC®

Supplier: MilliporeSigma
Warning

Synonyms: Ferrous sulphate hydrate, Ferrous sulfate hydrate

1.03967.9025
EM1.03967.9025EA 804.23 USD
EM1.03967.9025
Iron(II) sulfate hydrate 86-89% (by manganometry) FCC, BP, USP, SAFC®
Iron (II) sulphate hydrate
Formula: FeSO₄·H₂O
Melting Pt: ∼300 °C
Density: 2.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149722
CAS Number: 13463-43-9
EINECS: 231-753-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR