Ethyl acetate ≥99.8% (by GC), LiChrosolv®, hypergrade for LC-MS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Acetic acid ethyl ester, Acetic ether, Acetoxyethane, Acetidin, Ethyl acetic ester, Ethyl ethanoate, EtOAc, Vinegar naphtha, Acetic acid ethyl ester

M-Clarity™ quality level = MQ100

1.03649.2500
EM1.03649.2500EA 145 USD
EM1.03649.2500
Ethyl acetate ≥99.8% (by GC), LiChrosolv®, hypergrade for LC-MS, Supelco®
Ethyl acetate

LiChrosolv® hypergrade is accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionization methods (ESI/APCI - positive and negative mode).


Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionization efficiency. The packaging material has been improved to meet LC-MS quality requirements perfectly.

Formula: CH₃COOC₂H₅
Boiling Pt: 77 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.90 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥99.8%
Identity (IR) conforms
Evaporation Residue ≤2.0 mg/l
Water ≤0.05%
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g
Al (Aluminium) ≤10 ppb
Ca (Calcium) ≤10 ppb
Fe (Iron) ≤10 ppb
K (Potassium) ≤5 ppb
Mg (Magnesium) ≤10 ppb
Na (Sodium) ≤50 ppb
Transmission (at 260 nm) ≥50%
Transmission (at 265 nm) ≥80%
Transmission (from 270 nm) ≥98%
Suitable For LC-MS; Intensity Of Background Mass Peak Based On Reserpine (APCI/ESI Positive) ≤2 ppb
Suitable For LC-MS; Intensity Of Background Mass Peak Based On Reserpine (APCI/ESI Negative) ≤20 ppb
Filtered by 0.2 μm Filter


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR