Chloroform, GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

M-Clarity™ quality level = MQ300

1.02442.1000 1.02442.2500
EM1.02442.1000CS 644 USD
EM1.02442.1000 EM1.02442.2500
Chloroform, GR for analysis, Supelco®
Chloroform
Formula: CHCl₃
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR