tert-Butyl methyl ether ≥99% (by GC), EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: MTBE, 2-Methyl-2-methoxypropane, Ether, tert-butyl methyl, Methyl 1,1-dimethylethyl ether, 2-Methoxy-2-methylpropane, Methyl t-butyl ether, Methyl tert-butyl ether, Methyl-t-Butyl Ether

M-Clarity™ quality level = MQ200

1.01843.2500 1.01843.6025 1.01843.6190 1.01843.9011
EM1.01843.2500CS 698 USD
EM1.01843.2500 EM1.01843.6025 EM1.01843.6190 EM1.01843.9011
tert-Butyl methyl ether ≥99% (by GC), EMPLURA®, Supelco®
tert-Butyl methyl ether
Formula: (CH₃)₃COCH₃
Boiling Pt: 55.3 °C (1013 hPa)
Melting Pt: –108.6 °C
Density: 0.74 g/cm3 (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR