Ammonium sulfate ≥99.5% (by alkalimetry), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

1.01217.0100 1.01217.1000 1.01217.5000 1.01217.9025
EM1.01217.0100CS 6569 USD
EM1.01217.0100 EM1.01217.1000 EM1.01217.5000 EM1.01217.9025 EM1.01217.9050
Ammonium sulfate ≥99.5% (by alkalimetry), EMSURE® for analysis, Supelco®
Ammonium sulphate
Formula: (NH₄)₂SO₄
Melting Pt: >280 °C (dec., lit.)
Density: 1.77 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Solubility 754 g/l (20 °C)
Molar Mass 132.14 g/mol
Bulk Density 850 kg/m3
Density 1.77 g/cm3 (20 °C)
pH 5 (100 g/l, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR