Sulfuric acid 90-100%, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ300

1.00732.2510
EM1.00732.1000EA 106 USD
EM1.00732.1000 EM1.00732.2510
Sulfuric acid 90-100%, Supelco®
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84 g/cm³ (20 °C)
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR