Sulfuric acid 25% (by alkalimetry), Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.00716.1000 1.00716.9025
EM1.00716.1000CS 315 USD
EM1.00716.1000 EM1.00716.9025
Sulfuric acid 25% (by alkalimetry), Supelco®
Sulfuric acid

For general laboratory use.

Formula: H₂SO₄
Boiling Pt: ∼100 °C (1013 hPa)
Density: 1.1…1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Aluminum (Al) 0.05 ppm max.
Ammonium (NH4) 2 ppm max.
Arsenic (As) 0.01 ppm max.
Barium (Ba) 0.05 ppm max.
Beryllium (Be) 0.01 ppm max.
Bismuth (Bi) 0.1 ppm max.
Cadmium (Cd) 0.02 ppm max.
Calcium (Ca) 0.2 ppm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR