Hydrochloric acid 0.1000 M in 2-propanol, Titripur® DIN 51558-1, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

1.00326.1000
EM1.00326.1000CS 529 USD
EM1.00326.1000
Hydrochloric acid 0.1000 M in 2-propanol, Titripur® DIN 51558-1, Supelco®
Hydrochloric acid
Formula: HCl
Density: 0.79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3 (8),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Flash Point 12 °C (2-Propanol)
Explosion Limit 2 - 13.4 %(V) (2-Propanol)
Density 0.79 g/cm3 (20 °C)
pH  - 0.5 (H2O, 20 °C)
Ignition Point 425 °C (2-Propanol)
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR