Lactic acid ≥85% in aqueous solution, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

LX0020-3 LX0020-6
EM-LX0020-3EA 596 USD
EM-LX0020-3 EM-LX0020-6
Lactic acid ≥85% in aqueous solution, GR ACS, Supelco®
Lactic acid

Meets ACS Specifications

Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Density: 1.11…1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0
Merck Index: 13,05351

Order Now

Specification Test Results

Assay 85.0-90.0%
Chloride (Cl) 0.001% max
Color (APHA) 50 max
Form Liquid
Heavy metals (as Pb) 5ppm max
Iron (Fe) 5ppm max
Residue after ignition 0.02% max
Substances darkened by sulfuric acid To pass test
Sulfate (SO4) 0.002% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR