D-(+)-Glucose, anhydrous, GR ACS, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

DX0145-5 DX0145-1
EM-DX0145-5EA 526 USD
EM-DX0145-5 EM-DX0145-1
D-(+)-Glucose, anhydrous, GR ACS, Sigma-Aldrich®
D(+)-Glucose

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 150…152 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (description) powder
Appearance of solution passes test
Chloride (Cl) 0.01% max
Color White
Heavy metals (as Pb) 0.0005% max
Insoluble matter 0.005% max
Iron (Fe) 0.0005% max
Loss on drying 0.2% max
Sulfate and sulfite (as SO4) 0.005% max
Specific rotation [a]25/D +52.5-+53.0°
Starch passes test
Sulfated ash 0.02% max
Titrable acid 0.002meq/g max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR