Acetonitrile ≥99.5% (by GC), GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

AX0149-1 AX0149-6 AX0149-20 AX0149-3
EM-AX0149-1CS 2535 USD
EM-AX0149-1 EM-AX0149-6 EM-AX0149-20 EM-AX0149-3
Acetonitrile ≥99.5% (by GC), GR ACS, Supelco®
Acetonitrile

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81…82 °C (lit.)
Melting Pt: –45 °C (lit.)
Density: 0.786 g/ml (25°C) (lit.)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.5% min
Appearance Clear
Color (APHA) 10 max
Residue after evaporation 0.005% max
Titrable acid 8µeq/g max
Titrable base 0.6µeq/g max
Water (H2O) 0.3% max

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR