Polysorbate 20, OmniPur® for biochemistry DNase-, RNase-, protease-free, Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated

M-Clarity™ quality level = MQ100

9485-4L
EM-9485EA 588.15 USD
EM-9485 EM-9480
Polysorbate 20, OmniPur® for biochemistry DNase-, RNase-, protease-free, Calbiochem®, Millipore®
Polysorbate 20

Non-ionic detergent suited for solubilization of membrane proteins, enzymology, analytical, and diagnostic work, as well as chromatography and electrophoresis. Molecular Biology Grade.


Specifications
DNAse: none detected
Heavy metals (as Pb): ≤0.001%
Hydroxyl number: 96-108
Residue after ignition: ≤0.25%
Moisture (H2O): ≤3.0%
Protease: none detected
RNAse: none detected

Formula: C₅₈H₁₁₄O₂₆
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR