TAE Buffer, Concentrate, OmniPur*. 10X.
Supplier: MilliporeSigma
8720-4L
EM-8725EA 529.95 USD
EM-8725 EM-8720
TAE Buffer, Concentrate, OmniPur*. 10X.
Buffers Electrophoresis Buffers Running Buffers
Liquid.
  • DNase-, RNase-, and protease-free
Each liter of concentrate makes 10 L of 1X TAE (0.04 M TRIS, 0.04 M Acetate, 0.001 M EDTA).