50/84 Place Fusion Furnace, Furnace and Assay Supplies

Supplier: KLEN INTERNATIONAL (74) PTY LTD.
FAS-120-OIL FAS-120-NG1 FAS-120-LPG1 FAS-120-NG2 FAS-120-LPG2
CA11023-246EA 81809.6 USD
CA11023-246 CA11023-248 CA11023-250 CA76047-132 CA76047-134
50/84 Place Fusion Furnace, Furnace and Assay Supplies
Ovens Furnaces (Ashing and Muffle Ovens)
These 50/84-place fusion furnaces are available with LPG, natural gas, or diesel fuel source options

  • Suitable for large labs; 400-1200 samples per day including standards
  • Base construction uses high-quality 100 x 100 RHS
  • Furnace body construction is durable 5mm laser-cut plate
  • High quality refractory brick and insulation
  • Bolt holes in base plates of all 4 legs for individual adjustment on uneven surfaces
  • Pneumatic door controls with foot pedal controller—available with an optional hand valve controller

This furnace has a fan forced dual burner system,More Product Information
Order Now

SPECIFICATIONS

Shipping Dimensions 209 x 169 x 208 cm
Environmentally Preferable
Shipping Weight 2200 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR