Scorifier Steel Tongs, Webber's Welding
Supplier: Webber's Welding
FA118
CA11017-660EA 449.86 USD
CA11017-660
Scorifier Steel Tongs, Webber's Welding
Tongs
The scorifier tongs are made of mild steel for dependable, long-lasting use.
The longer length increases user safety, by distancing the user from the oven or furnace.
Ordering information: Contact your Avantor sales representative for more information.