Crucible Loading Fork, Webber's Welding
Supplier: Webber's Welding
FA114B FA114A
CA11017-642EA 449.86 USD
CA11017-642 CA11019-808 11019-810
Crucible Loading Fork, Webber's Welding
Forks
Crucible Fork is specially designed for heavy-duty applications with user safety in mind.
  • Aluminum pipe handle helps insulate against heat from ovens or furnaces
  • Mild steel crucible holders provide durable and dependable service
Ordering information: Overall Length: 50"
Handle Length: 46"
Order Now
SPECIFICATIONS
Environmentally Preferable