Aluminium ≥99.995% (trace metals basis), foil, Thickness 0.25 mm

Supplier: BeanTown Chemical

Synonyms: Aluminum

222630-100X100 222630-100X500 222630-50X50MM
BT222630-100X1EA 76 USD
BT222630-100X1 BT222630-100X5 BT222630-50X50
Aluminium ≥99.995% (trace metals basis), foil, Thickness 0.25 mm
Aluminium

CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029; RTECS: BD0330000 Solid; Molecular Formula: Al; MW: 26.98 Melting Point: 660.4°; Boiling Point: 2460° Density (g/mL): 2.7


  • RTECS: BD0330000
  • Solid

Formula: Al
Boiling Pt: 2460 °C
Melting Pt: 660.4 °C
Density: 2.7 g/mL
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR