(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%

Supplier: BeanTown Chemical

Synonyms: (R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

220380-1G 220380-250MG
BT220380-1GEA 110 USD
BT220380-1G BT220380-250MG
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

CAS: 76189-55-4; MDL No: MFCD00010805 Crystalline; Molecular Formula: C44H32P2; MW: 622.67 Melting Point: 239-241° Optical Rotation: [α]20/D +222°, c = 0.5% in benzene Air Sensitive


  • Optical rotation: [α]20/D +222°, c = 0.5% in benzene
  • Air Sensitive
  • Crystalline

Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
Melting Pt: 239…241 °C
MDL Number: MFCD00010805
CAS Number: 76189-55-4
Merck Index: 13,01223

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR