L(+)-Valine ≥99%

Supplier: BeanTown Chemical

Synonyms: H-Val-OH, Valine, (S)-(+)-Valine

218745-100G 218745-25G 218745-500G
BT218745-100GEA 67.52 USD
BT218745-100G BT218745-25G BT218745-500G
L(+)-Valine ≥99%
L(+)-Valine

CAS: 72-18-4; EC No: 200-773-6; MDL No: MFCD00064220; RTECS: YV9361000 Solid; Molecular Formula: C5H11NO2; MW: 117.15 Melting Point: 295-300° (sublimes) Optical Rotation: [α]20/D +27.0±0.5°, c = 5% in 5M HCl


  • RTECS: YV9361000
  • Optical rotation: [α]20/D +27.0±0.5°, c = 5% in 5M HCl
  • Solid

Formula: C₅H₁₁NO₂
Melting Pt: 295…300 °C (subl.)
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR