Isophthaloyl dichloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Isophthaloyl chloride

218625-100G 218625-500G
BT218625-100GEA 26.11 USD
BT218625-100G BT218625-500G
Isophthaloyl dichloride ≥98%
Isophthaloyl dichloride

CAS: 99-63-8; EC No: 202-774-7; MDL No: MFCD00000678; RTECS: NT2625000 UN No: UN3261; Haz Class: 8; Packing Group: II Solid; Molecular Formula: C8H4Cl2O2; MW: 203.02 Melting Point: 43-44°; Boiling Point: 276°; Flash point: 180°C (356°F) Moisture Sensitive


  • RTECS: NT2625000
  • Moisture Sensitive
  • Solid

Formula: C₈H₄Cl₂O₂
Boiling Pt: 276 °C
Melting Pt: 43…44 °C
Flash Pt: 180 °C (356 °F)
MDL Number: MFCD00000678
CAS Number: 99-63-8
EINECS: 202-774-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR