Calcium hydroxide ≥95.0% ACS

Supplier: BeanTown Chemical

Synonyms: Slaked lime

218185-1KG 218185-250G
BT218185-1KGEA 83.73 USD
BT218185-1KG BT218185-250G
Calcium hydroxide ≥95.0% ACS
Calcium hydroxide

CAS: 1305-62-0; EC No: 215-137-3; MDL No: MFCD00010901; RTECS: EW2800000 UN No: UN3262; Haz Class: 8; Packing Group: III Powder; Linear Formula: Ca(OH)2; MW: 74.09 Density (g/mL): 2.24 Air Sensitive


  • RTECS: EW2800000
  • Air Sensitive
  • Powder

Formula: Ca(OH)₂
Density: 2.24 g/mL
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3
UN: 3262
ADR: 8,III

Order Now

Specification Test Results

Assay 95.0% min.
Calcium carbonate ≤3.0%
Insoluble in HCl ≤0.03%
Chloride (Cl) ≤0.03%
Sulfur compounds (as SO4) ≤0.1%
Heavy metals (as Pb) ≤0.003%
Iron (Fe) ≤0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR