Titanium ≥99.995% (trace metals basis), slug, Length 6.35 mm, Diameter 3.175 mm

Supplier: BeanTown Chemical

217230-25G 217230-5G
BT217230-25GEA 159 USD
BT217230-25G BT217230-5G
Titanium ≥99.995% (trace metals basis), slug, Length 6.35 mm, Diameter 3.175 mm
Titanium

CAS: 7440-32-6; EC No: 231-142-3; MDL No: MFCD00011264 Solid; Molecular Formula: Ti; MW: 47.87 Melting Point: 1660°; Boiling Point: 3287° Density (g/mL): 4.5


  • Solid

Formula: Ti
Boiling Pt: 3287 °C
Melting Pt: 1660 °C
Density: 4.5 g/mL
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR