Creatine (N-amidinosarcosine) monohydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: Creatine Hydrate, 2-(1-Methylguanidino)acetic acid monohydrate, Creatine monohydrate, 2-(1-Methylguanidino)acetic acid hydrate, N-Amidinosarcosine monydrate

216960-100G 216960-2.5KG 216960-500G
BT216960-100GEA 46.87 USD
BT216960-100G BT216960-2.5KG BT216960-500G
Creatine (N-amidinosarcosine) monohydrate ≥99%
Creatine (N-Amidinosarcosine) monohydrate

CAS: 6020-87-7; EC No: 200-306-6; MDL No: MFCD00071582; RTECS: MB7706000 Powder; Molecular Formula: C4H9N3O2·H2O ; MW: 149.15


  • RTECS: MB7706000
  • Powder

Formula: C₄H₉N₃O₂·1H₂O
MW: 149.15 g/mol (131.13 anhy)
MDL Number: MFCD00071582
CAS Number: 6020-87-7
EINECS: 200-306-6

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR