Silver ≥99.9% (trace metals basis), foil, hard type, Thickness 0.28 mm

Supplier: BeanTown Chemical

212910-100X100 212910-100X500 212910-50X50MM
BT212910-100X1EA 287 USD
BT212910-100X1 BT212910-100X5 BT212910-50X50
Silver ≥99.9% (trace metals basis), foil, hard type, Thickness 0.28 mm
Silver

CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397 Solid; Molecular Formula: Ag; MW: 107.87 Melting Point: 960°; Boiling Point: 2212° Density (g/mL): 10.49


  • Solid

Formula: Ag
Boiling Pt: 2212 °C
Melting Pt: 960 °C
Density: 10.49 g/mL
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR