Sodium thiosulfate, anhydrous ≥99%

Supplier: BeanTown Chemical

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

144885-2.5KG 144885-500G
BT144885-2.5KGEA 126.24 USD
BT144885-2.5KG BT144885-500G
Sodium thiosulfate, anhydrous ≥99%
Sodium thiosulphate

CAS: 7772-98-7; EC No: 231-867-5; MDL No: MFCD00003499; RTECS: XN6476000 Powder; Linear Formula: Na2S2O3; MW: 158.11 Melting Point: 52° (decomposes) Density (g/mL): 1.667 Hygroscopic


  • RTECS: XN6476000
  • Hygroscopic
  • Powder

Formula: Na₂S₂O₃
Melting Pt: 52 °C (dec.)
Density: 1.667 g/mL
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR