L-Methionine ≥98%

Supplier: BeanTown Chemical

Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid

144115-100G 144115-2.5KG 144115-500G
BT144115-100GEA 65.79 USD
BT144115-100G BT144115-2.5KG BT144115-500G
L-Methionine ≥98%
L-Methionine

CAS: 63-68-3; EC No: 200-562-9; MDL No: MFCD00063097; RTECS: PD0457000 Powder; Molecular Formula: C5H11NO2S; MW: 149.21 Melting Point: 284° (decomposes) Optical Rotation: [α]20/D +23.7±0.5°, c = 5% in 5M HCl


  • RTECS: PD0457000
  • Optical rotation: [α]20/D +23.7±0.5°, c = 5% in 5M HCl
  • Powder

Formula: C₅H₁₁NO₂S
Melting Pt: 284 °C (dec.)
MDL Number: MFCD00063097
CAS Number: 63-68-3
EINECS: 200-562-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR